Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 179616 | |||||||||||||||||||||||
Literature Links: |
C11orf74 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
C11orf74 - chromosome 11 open reading frame 74 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAG2 - recombination activating gene 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000536.3 | 1683 | Missense Mutation | AAA,AGA | K,R 493 | NP_000527.2 | |
NM_001243785.1 | 1683 | Missense Mutation | AAA,AGA | K,R 493 | NP_001230714.1 | |
NM_001243786.1 | 1683 | Missense Mutation | AAA,AGA | K,R 493 | NP_001230715.1 |