Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCGACAATTAACCACTGGCATT[C/T]ATTTCCAAAGTATGCCGCAGTTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607957 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMK1D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAMK1D - calcium/calmodulin dependent protein kinase ID | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020397.3 | Intron | NP_065130.1 | ||||
NM_153498.3 | Intron | NP_705718.1 | ||||
XM_006717481.3 | Intron | XP_006717544.1 | ||||
XM_006717482.3 | Intron | XP_006717545.1 | ||||
XM_006717483.3 | Intron | XP_006717546.1 | ||||
XM_011519591.2 | Intron | XP_011517893.1 | ||||
XM_011519592.2 | Intron | XP_011517894.1 | ||||
XM_011519593.2 | Intron | XP_011517895.1 | ||||
XM_011519594.2 | Intron | XP_011517896.1 | ||||
XM_011519595.2 | Intron | XP_011517897.1 | ||||
XM_017016438.1 | Intron | XP_016871927.1 | ||||
XM_017016439.1 | Intron | XP_016871928.1 |