Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616049 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PABPC1L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PABPC1L - poly(A) binding protein cytoplasmic 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001124756.2 | 1742 | Intron | NP_001118228.1 |
TOMM34 - translocase of outer mitochondrial membrane 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006809.4 | 1742 | UTR 3 | NP_006800.2 | |||
XM_005260254.2 | 1742 | UTR 3 | XP_005260311.1 | |||
XM_011528501.1 | 1742 | UTR 3 | XP_011526803.1 | |||
XM_017027600.1 | 1742 | UTR 3 | XP_016883089.1 |