Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613362 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CINP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CINP - cyclin dependent kinase 2 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320046.1 | 1804 | Intron | NP_001306975.1 | |||
NM_032630.2 | 1804 | Intron | NP_116019.1 |
ZNF839 - zinc finger protein 839 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267827.1 | 1804 | Missense Mutation | CCA,TCA | P,S 531 | NP_001254756.1 | |
NM_001267828.1 | 1804 | Missense Mutation | CCA,TCA | P,S 531 | NP_001254757.1 | |
NM_018335.4 | 1804 | Missense Mutation | CCA,TCA | P,S 647 | NP_060805.3 | |
XM_006720203.3 | 1804 | Missense Mutation | CCA,TCA | P,S 559 | XP_006720266.2 | |
XM_011536945.1 | 1804 | Missense Mutation | CCA,TCA | P,S 681 | XP_011535247.1 | |
XM_011536946.2 | 1804 | Missense Mutation | CCA,TCA | P,S 631 | XP_011535248.1 | |
XM_011536948.2 | 1804 | Missense Mutation | CCA,TCA | P,S 531 | XP_011535250.1 | |
XM_011536949.2 | 1804 | Intron | XP_011535251.1 | |||
XM_011536950.2 | 1804 | Intron | XP_011535252.1 | |||
XM_017021447.1 | 1804 | Missense Mutation | CCA,TCA | P,S 597 | XP_016876936.1 | |
XM_017021448.1 | 1804 | Intron | XP_016876937.1 | |||
XM_017021449.1 | 1804 | Missense Mutation | CCA,TCA | P,S 312 | XP_016876938.1 | |
XM_017021450.1 | 1804 | Intron | XP_016876939.1 | |||
XM_017021451.1 | 1804 | Intron | XP_016876940.1 |