Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613842 MIM: 611270 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GZF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GZF1 - GDNF inducible zinc finger protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317012.1 | 3616 | Intron | NP_001303941.1 | |||
NM_001317019.1 | 3616 | Intron | NP_001303948.1 | |||
NM_022482.4 | 3616 | Intron | NP_071927.1 |
NAPB - NSF attachment protein beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283018.1 | 3616 | UTR 3 | NP_001269947.1 | |||
NM_001283020.1 | 3616 | UTR 3 | NP_001269949.1 | |||
NM_001283026.1 | 3616 | UTR 3 | NP_001269955.1 | |||
NM_022080.2 | 3616 | UTR 3 | NP_071363.1 | |||
XM_011529313.1 | 3616 | UTR 3 | XP_011527615.1 | |||
XM_011529315.2 | 3616 | Intron | XP_011527617.1 | |||
XM_017028008.1 | 3616 | UTR 3 | XP_016883497.1 |
Set Membership: |
HapMap |