Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCTTTTTCACCGCCAGGGACCC[C/T]GGAGTGGAGATGACAGTGTTCCGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600050 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP3K11 - mitogen-activated protein kinase kinase kinase 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCNX3 - pecanex homolog 3 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032223.3 | 2232 | Silent Mutation | CCC,CCT | P,P 116 | NP_115599.2 | |
XM_005273987.4 | 2232 | Silent Mutation | CCC,CCT | P,P 116 | XP_005274044.1 | |
XM_005273990.4 | 2232 | Silent Mutation | CCC,CCT | P,P 116 | XP_005274047.1 | |
XM_006718549.3 | 2232 | Silent Mutation | CCC,CCT | P,P 116 | XP_006718612.1 | |
XM_006718551.3 | 2232 | Silent Mutation | CCC,CCT | P,P 116 | XP_006718614.1 | |
XM_011545024.2 | 2232 | Silent Mutation | CCC,CCT | P,P 116 | XP_011543326.1 | |
XM_011545025.2 | 2232 | Silent Mutation | CCC,CCT | P,P 116 | XP_011543327.1 | |
XM_011545026.2 | 2232 | Intron | XP_011543328.1 | |||
XM_011545027.1 | 2232 | Intron | XP_011543329.1 | |||
XM_011545028.2 | 2232 | Silent Mutation | CCC,CCT | P,P 116 | XP_011543330.1 |