Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610952 MIM: 610833 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRNKL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CRNKL1 - crooked neck pre-mRNA splicing factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAA20 - N(alpha)-acetyltransferase 20, NatB catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016100.4 | 233 | Silent Mutation | TAC,TAT | Y,Y 42 | NP_057184.1 | |
NM_181527.3 | 233 | Silent Mutation | TAC,TAT | Y,Y 30 | NP_852668.1 | |
NM_181528.3 | 233 | Silent Mutation | TAC,TAT | Y,Y 42 | NP_852669.1 |
Set Membership: |
HapMap Validated |