Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615677 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERPINA12 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SERPINA12 - serpin family A member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304461.1 | Intron | NP_001291390.1 | ||||
NM_173850.3 | Intron | NP_776249.1 | ||||
XM_011536451.2 | Intron | XP_011534753.1 | ||||
XM_011536452.2 | Intron | XP_011534754.1 | ||||
XM_011536453.2 | Intron | XP_011534755.1 | ||||
XM_011536454.2 | Intron | XP_011534756.1 | ||||
XM_011536455.1 | Intron | XP_011534757.1 | ||||
XM_017020989.1 | Intron | XP_016876478.1 | ||||
XM_017020990.1 | Intron | XP_016876479.1 |
SERPINA9 - serpin family A member 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |