Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGACTACCAGAAAATCACCTCC[G/A]TGCTCTTTCAAGCCCTCGGATCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602110 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102724064 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC102724064 - uncharacterized LOC102724064 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC29A2 - solute carrier family 29 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300868.1 | Intron | NP_001287797.1 | ||||
NM_001300869.1 | Intron | NP_001287798.1 | ||||
NM_001532.2 | Intron | NP_001523.2 | ||||
XM_011544974.1 | Intron | XP_011543276.1 | ||||
XM_017017630.1 | Intron | XP_016873119.1 | ||||
XM_017017631.1 | Intron | XP_016873120.1 | ||||
XM_017017632.1 | Intron | XP_016873121.1 | ||||
XM_017017633.1 | Intron | XP_016873122.1 | ||||
XM_017017634.1 | Intron | XP_016873123.1 | ||||
XM_017017635.1 | Intron | XP_016873124.1 | ||||
XM_017017636.1 | Intron | XP_016873125.1 | ||||
XM_017017637.1 | Intron | XP_016873126.1 | ||||
XM_017017638.1 | Intron | XP_016873127.1 |
Set Membership: |
HapMap Validated |