Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611510 MIM: 607720 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CENPT - centromere protein T | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025082.3 | Intron | NP_079358.3 | ||||
XM_006721277.2 | Intron | XP_006721340.1 | ||||
XM_011523344.1 | Intron | XP_011521646.1 | ||||
XM_011523345.1 | Intron | XP_011521647.1 | ||||
XM_017023712.1 | Intron | XP_016879201.1 | ||||
XM_017023713.1 | Intron | XP_016879202.1 | ||||
XM_017023714.1 | Intron | XP_016879203.1 | ||||
XM_017023715.1 | Intron | XP_016879204.1 | ||||
XM_017023716.1 | Intron | XP_016879205.1 | ||||
XM_017023717.1 | Intron | XP_016879206.1 |
TSNAXIP1 - translin associated factor X interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |