Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602659 | ||||||||||||||||||||
Literature Links: |
MNAT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MNAT1 - MNAT1, CDK activating kinase assembly factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177963.1 | 183 | Silent Mutation | CTG,TTG | L,L 19 | NP_001171434.1 | |
NM_002431.3 | 183 | Silent Mutation | CTG,TTG | L,L 19 | NP_002422.1 | |
XM_005267687.2 | 183 | Silent Mutation | CTG,TTG | L,L 19 | XP_005267744.1 | |
XM_005267688.2 | 183 | Silent Mutation | CTG,TTG | L,L 19 | XP_005267745.1 | |
XM_017021332.1 | 183 | Intron | XP_016876821.1 | |||
XM_017021333.1 | 183 | UTR 5 | XP_016876822.1 | |||
XM_017021334.1 | 183 | Intron | XP_016876823.1 |