Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAACACTGAGTCTGTAAATCTACA[G/T]ACATACAATATTCGTTGAAGTCTAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616967 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALOX15P1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ALOX15P1 - arachidonate 15-lipoxygenase pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C17orf100 - chromosome 17 open reading frame 100 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIAA0753 - KIAA0753 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MED31 - mediator complex subunit 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016060.2 | Intron | NP_057144.1 | ||||
XM_017024710.1 | Intron | XP_016880199.1 |
MIR4520-1 - microRNA 4520-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4520-2 - microRNA 4520-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TXNDC17 - thioredoxin domain containing 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |