Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612687 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGMB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RGMB - repulsive guidance molecule family member b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012761.2 | Intron | NP_001012779.2 | ||||
XM_005271966.3 | Intron | XP_005272023.1 | ||||
XM_011543345.2 | Intron | XP_011541647.1 | ||||
XM_011543346.2 | Intron | XP_011541648.1 | ||||
XM_011543347.2 | Intron | XP_011541649.2 | ||||
XM_017009390.1 | Intron | XP_016864879.1 | ||||
XM_017009391.1 | Intron | XP_016864880.1 |
RGMB-AS1 - RGMB antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |