Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGATGAAGCGGGAGATGATGGGG[G/C]GCCGCAGCAGGTTCTGAACCGTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300203 MIM: 300839 | ||||||||||||||||||||
Literature Links: |
CDKL5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDKL5 - cyclin dependent kinase like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037343.1 | 610 | Intron | NP_001032420.1 | |||
NM_001323289.1 | 610 | Intron | NP_001310218.1 | |||
NM_003159.2 | 610 | Intron | NP_003150.1 |
RS1 - retinoschisin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000330.3 | 610 | Missense Mutation | CCC,CGC | P,R 192 | NP_000321.1 |