Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607566 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPM2A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EPM2A - epilepsy, progressive myoclonus type 2A, Lafora disease (laforin) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018041.1 | Intron | NP_001018051.1 | ||||
NM_005670.3 | Intron | NP_005661.1 | ||||
XM_006715564.3 | Intron | XP_006715627.1 | ||||
XM_011536113.2 | Intron | XP_011534415.1 | ||||
XM_011536116.1 | Intron | XP_011534418.1 | ||||
XM_017011300.1 | Intron | XP_016866789.1 | ||||
XM_017011301.1 | Intron | XP_016866790.1 | ||||
XM_017011302.1 | Intron | XP_016866791.1 |
Set Membership: |
HapMap |