Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTACTATTTAACCCAAACATAC[A/C]TTTTTAATACACAATTTGTACCTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611681 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS20 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ADAMTS20 - ADAM metallopeptidase with thrombospondin type 1 motif 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025003.3 | 6417 | Intron | NP_079279.3 | |||
XM_011538754.2 | 6417 | UTR 3 | XP_011537056.1 | |||
XM_017019979.1 | 6417 | UTR 3 | XP_016875468.1 |
Set Membership: |
HapMap |