Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107470 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFNGR1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IFNGR1 - interferon gamma receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000416.2 | 1574 | Missense Mutation | CCT,CTT | P,L 467 | NP_000407.1 | |
XM_006715470.2 | 1574 | Missense Mutation | CCT,CTT | P,L 457 | XP_006715533.1 | |
XM_006715471.2 | 1574 | Missense Mutation | CCT,CTT | P,L 426 | XP_006715534.1 | |
XM_011535793.2 | 1574 | Missense Mutation | CCT,CTT | P,L 457 | XP_011534095.1 | |
XM_011535794.1 | 1574 | Missense Mutation | CCT,CTT | P,L 457 | XP_011534096.1 | |
XM_017010827.1 | 1574 | Missense Mutation | CCT,CTT | P,L 457 | XP_016866316.1 |
Set Membership: |
HapMap |