Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGAAAATAAAATCCCATAACAA[C/T]GGCAACAAATAATCCAGATTCAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105372269 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC105372269 - endogenous retrovirus group K member 8 Gag polyprotein-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011528461.2 | Intron | XP_011526763.1 |
ZNF560 - zinc finger protein 560 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152476.2 | Intron | NP_689689.2 | ||||
XM_011527696.2 | Intron | XP_011525998.1 | ||||
XM_011527697.2 | Intron | XP_011525999.1 | ||||
XM_017026327.1 | Intron | XP_016881816.1 | ||||
XM_017026328.1 | Intron | XP_016881817.1 | ||||
XM_017026329.1 | Intron | XP_016881818.1 | ||||
XM_017026330.1 | Intron | XP_016881819.1 |
Set Membership: |
HapMap Validated |