Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607815 MIM: 602233 | ||||||||||||||||||||
Literature Links: |
ANKS1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APAF1 - apoptotic peptidase activating factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160.2 | 4373 | UTR 3 | NP_001151.1 | |||
NM_013229.2 | 4373 | UTR 3 | NP_037361.1 | |||
NM_181861.1 | 4373 | UTR 3 | NP_863651.1 | |||
NM_181868.1 | 4373 | UTR 3 | NP_863658.1 | |||
NM_181869.1 | 4373 | UTR 3 | NP_863659.1 | |||
XM_017019250.1 | 4373 | Intron | XP_016874739.1 |