Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTAGTGATAGCTGGAAGGAAAAA[C/G]TGTCCCCTCAATAGAAATAATGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608846 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADM5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ADM5 - adrenomedullin 5 (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CPT1C - carnitine palmitoyltransferase 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136052.2 | Intron | NP_001129524.1 | ||||
NM_001199752.2 | Intron | NP_001186681.1 | ||||
NM_001199753.1 | Intron | NP_001186682.1 | ||||
NM_152359.2 | Intron | NP_689572.1 | ||||
XM_005258505.2 | Intron | XP_005258562.1 | ||||
XM_005258506.4 | Intron | XP_005258563.1 | ||||
XM_006723009.3 | Intron | XP_006723072.1 | ||||
XM_011526438.2 | Intron | XP_011524740.1 | ||||
XM_011526439.2 | Intron | XP_011524741.1 | ||||
XM_011526440.2 | Intron | XP_011524742.1 | ||||
XM_017026265.1 | Intron | XP_016881754.1 | ||||
XM_017026266.1 | Intron | XP_016881755.1 | ||||
XM_017026267.1 | Intron | XP_016881756.1 | ||||
XM_017026268.1 | Intron | XP_016881757.1 | ||||
XM_017026269.1 | Intron | XP_016881758.1 | ||||
XM_017026270.1 | Intron | XP_016881759.1 | ||||
XM_017026271.1 | Intron | XP_016881760.1 | ||||
XM_017026272.1 | Intron | XP_016881761.1 | ||||
XM_017026273.1 | Intron | XP_016881762.1 |
Set Membership: |
HapMap |