Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTATAAACGTGCATGTACAAACAC[C/T]TTTTTCAGCGTTGGGACTAAATTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF233 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF233 - zinc finger protein 233 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207005.1 | Intron | NP_001193934.1 | ||||
NM_181756.2 | Intron | NP_861421.2 | ||||
XM_017026758.1 | Intron | XP_016882247.1 | ||||
XM_017026759.1 | Intron | XP_016882248.1 | ||||
XM_017026760.1 | Intron | XP_016882249.1 | ||||
XM_017026761.1 | Intron | XP_016882250.1 |
Set Membership: |
HapMap |