Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605776 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGL1 - fibrinogen like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004467.3 | 1165 | UTR 3 | NP_004458.3 | |||
NM_147203.2 | 1165 | UTR 3 | NP_671736.2 | |||
NM_201552.1 | 1165 | UTR 3 | NP_963846.1 | |||
NM_201553.1 | 1165 | UTR 3 | NP_963847.1 | |||
XM_017013232.1 | 1165 | UTR 3 | XP_016868721.1 | |||
XM_017013233.1 | 1165 | UTR 3 | XP_016868722.1 |