Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610308 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
B3GLCT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
B3GLCT - beta 3-glucosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_194318.3 | 3744 | UTR 3 | NP_919299.3 | |||
XM_006719768.3 | 3744 | UTR 3 | XP_006719831.1 | |||
XM_011534936.1 | 3744 | UTR 3 | XP_011533238.1 | |||
XM_011534938.2 | 3744 | UTR 3 | XP_011533240.1 | |||
XM_017020395.1 | 3744 | UTR 3 | XP_016875884.1 |
Set Membership: |
HapMap |