Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTACATGTTGTCATGTTAGTTAAC[A/G]TGGCTAGCTTGCCCTTTGACATCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF845 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF845 - zinc finger protein 845 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321522.1 | Intron | NP_001308451.1 | ||||
NM_001321523.1 | Intron | NP_001308452.1 | ||||
NM_001321524.1 | Intron | NP_001308453.1 | ||||
NM_138374.2 | Intron | NP_612383.1 |