Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAATAATTACATTATTTTTAGGAA[A/C]TCTTATCAAATATGGTCCTGGATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604001 MIM: 601637 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AKAP9 - A-kinase anchoring protein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005751.4 | Intron | NP_005742.4 | ||||
NM_147185.2 | Intron | NP_671714.1 |
CYP51A1 - cytochrome P450 family 51 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000786.3 | Intron | NP_000777.1 | ||||
NM_001146152.1 | Intron | NP_001139624.1 |
Set Membership: |
DME Validated Inventoried |