Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603709 MIM: 182520 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM22 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
ADAM22 - ADAM metallopeptidase domain 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRI - sorcin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256891.1 | 763 | Intron | NP_001243820.1 | |||
NM_001256892.1 | 763 | Intron | NP_001243821.1 | |||
NM_003130.3 | 763 | UTR 3 | NP_003121.1 | |||
NM_198901.1 | 763 | UTR 3 | NP_944490.1 | |||
XM_011516528.1 | 763 | UTR 3 | XP_011514830.1 |
Set Membership: |
HapMap Validated |