Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603785 | ||||||||||||||||||||
Literature Links: |
MPDZ PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MPDZ - multiple PDZ domain crumbs cell polarity complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261406.1 | 6224 | Missense Mutation | CAT,CGT | H,R 1997 | NP_001248335.1 | |
NM_001261407.1 | 6224 | Missense Mutation | CAT,CGT | H,R 1968 | NP_001248336.1 | |
NM_003829.4 | 6224 | Missense Mutation | CAT,CGT | H,R 2001 | NP_003820.2 | |
XM_005251622.4 | 6224 | Missense Mutation | CAT,CGT | H,R 2030 | XP_005251679.1 | |
XM_005251623.4 | 6224 | Missense Mutation | CAT,CGT | H,R 1960 | XP_005251680.1 | |
XM_006716885.3 | 6224 | Missense Mutation | CAT,CGT | H,R 2030 | XP_006716948.1 | |
XM_006716886.3 | 6224 | Missense Mutation | CAT,CGT | H,R 2030 | XP_006716949.1 | |
XM_006716887.3 | 6224 | Missense Mutation | CAT,CGT | H,R 2030 | XP_006716950.1 | |
XM_006716888.3 | 6224 | Missense Mutation | CAT,CGT | H,R 2001 | XP_006716951.1 | |
XM_006716889.3 | 6224 | Missense Mutation | CAT,CGT | H,R 1997 | XP_006716952.1 | |
XM_006716891.3 | 6224 | Missense Mutation | CAT,CGT | H,R 1968 | XP_006716954.1 | |
XM_017015252.1 | 6224 | Missense Mutation | CAT,CGT | H,R 2002 | XP_016870741.1 | |
XM_017015253.1 | 6224 | Missense Mutation | CAT,CGT | H,R 1968 | XP_016870742.1 | |
XM_017015254.1 | 6224 | Missense Mutation | CAT,CGT | H,R 1960 | XP_016870743.1 | |
XM_017015255.1 | 6224 | Missense Mutation | CAT,CGT | H,R 1960 | XP_016870744.1 | |
XM_017015256.1 | 6224 | Missense Mutation | CAT,CGT | H,R 1931 | XP_016870745.1 | |
XM_017015257.1 | 6224 | Missense Mutation | CAT,CGT | H,R 1931 | XP_016870746.1 | |
XM_017015258.1 | 6224 | Intron | XP_016870747.1 | |||
XM_017015259.1 | 6224 | Intron | XP_016870748.1 |