Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGTGGGCTACTCTCAGAATTTG[C/T]CTTCAGTTCTCTTAAAAAGGGGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610190 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHST8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHST8 - carbohydrate sulfotransferase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127895.1 | Intron | NP_001121367.1 | ||||
NM_001127896.1 | Intron | NP_001121368.1 | ||||
NM_022467.3 | Intron | NP_071912.2 | ||||
XM_011527222.1 | Intron | XP_011525524.1 | ||||
XM_011527224.1 | Intron | XP_011525526.1 | ||||
XM_017027143.1 | Intron | XP_016882632.1 |