Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605496 MIM: 612673 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNTRL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CNTRL - centriolin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007018.4 | 3982 | Intron | NP_008949.4 | |||
XM_005251679.3 | 3982 | Intron | XP_005251736.1 | |||
XM_006716940.2 | 3982 | Intron | XP_006717003.1 | |||
XM_006716942.1 | 3982 | Intron | XP_006717005.1 | |||
XM_006716944.3 | 3982 | Intron | XP_006717007.1 | |||
XM_011518166.1 | 3982 | Intron | XP_011516468.1 | |||
XM_011518167.1 | 3982 | Intron | XP_011516469.1 | |||
XM_011518168.1 | 3982 | Intron | XP_011516470.1 | |||
XM_011518169.2 | 3982 | Intron | XP_011516471.1 | |||
XM_011518170.1 | 3982 | Intron | XP_011516472.1 | |||
XM_011518172.2 | 3982 | Intron | XP_011516474.1 | |||
XM_011518173.1 | 3982 | Intron | XP_011516475.1 | |||
XM_017014223.1 | 3982 | Intron | XP_016869712.1 | |||
XM_017014224.1 | 3982 | Intron | XP_016869713.1 | |||
XM_017014225.1 | 3982 | Intron | XP_016869714.1 | |||
XM_017014226.1 | 3982 | Intron | XP_016869715.1 | |||
XM_017014227.1 | 3982 | Intron | XP_016869716.1 | |||
XM_017014228.1 | 3982 | Intron | XP_016869717.1 | |||
XM_017014229.1 | 3982 | Intron | XP_016869718.1 | |||
XM_017014230.1 | 3982 | Intron | XP_016869719.1 |
RAB14 - RAB14, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016322.3 | 3982 | UTR 3 | NP_057406.2 |