Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTCCTTCTTTTTTTTTCCTGAT[C/T]CATTATATTTATACGTATTCATGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612711 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GCC2 - GRIP and coiled-coil domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181453.3 | Intron | NP_852118.1 | ||||
XM_006712870.3 | Intron | XP_006712933.1 | ||||
XM_006712871.1 | Intron | XP_006712934.1 | ||||
XM_006712872.3 | Intron | XP_006712935.1 | ||||
XM_011512213.2 | Intron | XP_011510515.1 |
Set Membership: |
HapMap |