Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCAGGTACAGTGCATTGCTCCA[A/G]GTGTCATGCAAAGCTAGAAAGTCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 102681 | ||||||||||||||||||||
Literature Links: |
ADD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADD2 - adducin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185054.1 | 3353 | UTR 3 | NP_001171983.1 | |||
NM_001185055.1 | 3353 | Intron | NP_001171984.1 | |||
NM_001617.3 | 3353 | UTR 3 | NP_001608.1 | |||
NM_017482.3 | 3353 | Intron | NP_059516.2 | |||
NM_017488.3 | 3353 | UTR 3 | NP_059522.1 | |||
XM_011532502.2 | 3353 | UTR 3 | XP_011530804.1 |