Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTGCTGATGGGGGCTCCTCCGG[G/T]CTGAGGCACAGGGTGGGGAGGTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMP8A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BMP8A - bone morphogenetic protein 8a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181809.3 | Intron | NP_861525.2 | ||||
XM_006710616.3 | Intron | XP_006710679.1 | ||||
XM_011541381.2 | Intron | XP_011539683.1 | ||||
XM_011541382.2 | Intron | XP_011539684.1 | ||||
XM_017001198.1 | Intron | XP_016856687.1 |
LOC105378950 - mitogen-activated protein kinase 7-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |