Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610715 | ||||||||||||||||||||
Literature Links: |
HEMGN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HEMGN - hemogen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018437.4 | 1412 | Missense Mutation | AAG,ACG | K,T 416 | NP_060907.2 | |
NM_197978.2 | 1412 | Missense Mutation | AAG,ACG | K,T 416 | NP_932095.1 | |
XM_005252086.1 | 1412 | Missense Mutation | AAG,ACG | K,T 416 | XP_005252143.1 | |
XM_011518845.2 | 1412 | Missense Mutation | AAG,ACG | K,T 416 | XP_011517147.1 | |
XM_011518846.2 | 1412 | Missense Mutation | AAG,ACG | K,T 416 | XP_011517148.1 |
TRMO - tRNA methyltransferase O | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |