Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603136 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CUL3 - cullin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257197.1 | 3220 | UTR 3 | NP_001244126.1 | |||
NM_001257198.1 | 3220 | UTR 3 | NP_001244127.1 | |||
NM_003590.4 | 3220 | UTR 3 | NP_003581.1 | |||
XM_006712800.2 | 3220 | UTR 3 | XP_006712863.2 | |||
XM_011511994.2 | 3220 | UTR 3 | XP_011510296.1 | |||
XM_011511995.1 | 3220 | UTR 3 | XP_011510297.1 | |||
XM_011511996.2 | 3220 | UTR 3 | XP_011510298.1 |
Set Membership: |
HapMap Validated |