Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604683 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF3A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KIF3A - kinesin family member 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300791.1 | Intron | NP_001287720.1 | ||||
NM_001300792.1 | Intron | NP_001287721.1 | ||||
NM_007054.6 | Intron | NP_008985.3 | ||||
XM_006714526.3 | Intron | XP_006714589.1 | ||||
XM_017008995.1 | Intron | XP_016864484.1 | ||||
XM_017008996.1 | Intron | XP_016864485.1 |
Set Membership: |
HapMap |