Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601496 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GFRA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GFRA1 - GDNF family receptor alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145453.2 | 7177 | UTR 3 | NP_001138925.1 | |||
NM_005264.5 | 7177 | UTR 3 | NP_005255.1 | |||
NM_145793.3 | 7177 | UTR 3 | NP_665736.1 | |||
XM_005269696.1 | 7177 | UTR 3 | XP_005269753.1 | |||
XM_011539634.1 | 7177 | UTR 3 | XP_011537936.1 |
Set Membership: |
HapMap |