Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607422 MIM: 142708 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCAT PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GCAT - glycine C-acetyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171690.1 | Intron | NP_001165161.1 | ||||
NM_014291.3 | Intron | NP_055106.1 | ||||
XM_005261409.3 | Intron | XP_005261466.1 | ||||
XM_005261410.3 | Intron | XP_005261467.1 | ||||
XM_005261411.1 | Intron | XP_005261468.1 | ||||
XM_011530023.1 | Intron | XP_011528325.1 | ||||
XM_017028674.1 | Intron | XP_016884163.1 | ||||
XM_017028675.1 | Intron | XP_016884164.1 | ||||
XM_017028676.1 | Intron | XP_016884165.1 | ||||
XM_017028677.1 | Intron | XP_016884166.1 |
H1F0 - H1 histone family member 0 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |