Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGCTTCAACAATTCTTTATATC[A/G]TATTTTATTCTTGAAATTGGTATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SESTD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SESTD1 - SEC14 and spectrin domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178123.4 | 6419 | UTR 3 | NP_835224.3 | |||
XM_011512139.2 | 6419 | UTR 3 | XP_011510441.1 | |||
XM_011512141.2 | 6419 | UTR 3 | XP_011510443.1 | |||
XM_017005250.1 | 6419 | UTR 3 | XP_016860739.1 | |||
XM_017005251.1 | 6419 | UTR 3 | XP_016860740.1 |