Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603309 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK13 - cyclin dependent kinase 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003718.4 | 62 | Intron | NP_003709.3 | |||
NM_031267.3 | 62 | Intron | NP_112557.2 | |||
XM_011515597.2 | 62 | UTR 5 | XP_011513899.1 | |||
XM_017012750.1 | 62 | UTR 5 | XP_016868239.1 | |||
XM_017012751.1 | 62 | UTR 5 | XP_016868240.1 |