Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTATCACCCAAAGGGTCTCAGGA[A/G]CTTGAAAGAGGGACTTGAATCAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602642 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNFSF11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TNFSF11 - tumor necrosis factor superfamily member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003701.3 | Intron | NP_003692.1 | ||||
NM_033012.3 | Intron | NP_143026.1 | ||||
XM_011535280.2 | Intron | XP_011533582.1 | ||||
XM_017020802.1 | Intron | XP_016876291.1 | ||||
XM_017020803.1 | Intron | XP_016876292.1 |