Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBC1D21 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TBC1D21 - TBC1 domain family member 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286434.1 | Intron | NP_001273363.1 | ||||
NM_153356.2 | Intron | NP_699187.1 | ||||
XM_006720409.3 | Intron | XP_006720472.1 | ||||
XM_006720410.3 | Intron | XP_006720473.1 | ||||
XM_006720411.3 | Intron | XP_006720474.1 | ||||
XM_011521281.2 | Intron | XP_011519583.1 | ||||
XM_011521282.2 | Intron | XP_011519584.1 | ||||
XM_011521283.2 | Intron | XP_011519585.1 | ||||
XM_011521284.2 | Intron | XP_011519586.1 |
Set Membership: |
HapMap Validated |