Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147781 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL4R PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IL4R - interleukin 4 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000418.3 | Intron | NP_000409.1 | ||||
NM_001257406.1 | Intron | NP_001244335.1 | ||||
NM_001257407.1 | Intron | NP_001244336.1 | ||||
NM_001257997.1 | Intron | NP_001244926.1 | ||||
XM_005255308.2 | Intron | XP_005255365.1 | ||||
XM_006721043.1 | Intron | XP_006721106.1 | ||||
XM_011545825.1 | Intron | XP_011544127.1 | ||||
XM_011545826.2 | Intron | XP_011544128.1 | ||||
XM_011545827.2 | Intron | XP_011544129.1 | ||||
XM_011545828.2 | Intron | XP_011544130.1 | ||||
XM_011545829.1 | Intron | XP_011544131.1 | ||||
XM_011545830.2 | Intron | XP_011544132.1 | ||||
XM_011545831.1 | Intron | XP_011544133.1 | ||||
XM_011545832.1 | Intron | XP_011544134.1 | ||||
XM_011545833.2 | Intron | XP_011544135.1 | ||||
XM_011545834.2 | Intron | XP_011544136.1 | ||||
XM_017023211.1 | Intron | XP_016878700.1 |
Set Membership: |
HapMap |