Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACACAAATGACAGAAAACAACTA[C/T]GCATTATGGAACCTTTATTTTTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164360 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATP5A1 - ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1, cardiac muscle | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001935.2 | 1917 | UTR 3 | NP_001001935.1 | |||
NM_001001937.1 | 1917 | UTR 3 | NP_001001937.1 | |||
NM_001257334.1 | 1917 | UTR 3 | NP_001244263.1 | |||
NM_001257335.1 | 1917 | UTR 3 | NP_001244264.1 | |||
NM_004046.5 | 1917 | UTR 3 | NP_004037.1 | |||
XM_017025789.1 | 1917 | UTR 3 | XP_016881278.1 |
Set Membership: |
HapMap |