Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTCTTTTAAAATAAGTTCACTTC[A/G]GATTTTATTAGTTTGCCTCTTATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610132 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VANGL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VANGL1 - VANGL planar cell polarity protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172411.1 | Intron | NP_001165882.1 | ||||
NM_001172412.1 | Intron | NP_001165883.1 | ||||
NM_138959.2 | Intron | NP_620409.1 |