Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTCTGGCCCCTGCGTGGCCAGCA[G/A]GGCCGGCTCCTCCGGAGGCTCCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC101929060 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101929060 - uncharacterized LOC101929060 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIER2 - MIER family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005259582.4 | 3017 | UTR 3 | XP_005259639.1 | |||
XM_006722769.3 | 3017 | UTR 3 | XP_006722832.1 | |||
XM_011528072.2 | 3017 | UTR 3 | XP_011526374.1 | |||
XM_011528073.1 | 3017 | UTR 3 | XP_011526375.1 | |||
XM_011528074.2 | 3017 | UTR 3 | XP_011526376.1 | |||
XM_011528075.2 | 3017 | UTR 3 | XP_011526377.1 | |||
XM_011528077.2 | 3017 | UTR 3 | XP_011526379.1 | |||
XM_011528081.2 | 3017 | UTR 3 | XP_011526383.1 | |||
XM_011528082.2 | 3017 | UTR 3 | XP_011526384.1 |