Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGGGCAAGAGAACTAGATTTCA[G/T]GTATTGATTGTGGTCTTTGTTCAGT
Species: |
Human | |||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609234 | |||||||||||||||||||||||||||||||||||
Literature Links: |
EIF2A PubMed Links | |||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | ||||||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR - Not Available | ||||||||
AMR - Not Available |
EIF2A - eukaryotic translation initiation factor 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319043.1 | Intron | NP_001305972.1 | ||||
NM_001319044.1 | Intron | NP_001305973.1 | ||||
NM_001319045.1 | Intron | NP_001305974.1 | ||||
NM_001319046.1 | Intron | NP_001305975.1 | ||||
NM_032025.4 | Intron | NP_114414.2 | ||||
XM_011513224.1 | Intron | XP_011511526.1 |
SERP1 - stress-associated endoplasmic reticulum protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014445.3 | Intron | NP_055260.1 | ||||
XM_011512698.2 | Intron | XP_011511000.1 |
Set Membership: |
Validated |