Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603639 MIM: 606029 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM17 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ADAM17 - ADAM metallopeptidase domain 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CPSF3 - cleavage and polyadenylation specific factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IAH1 - isoamyl acetate-hydrolyzing esterase 1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039613.2 | Intron | NP_001034702.1 | ||||
NM_001320858.1 | Intron | NP_001307787.1 | ||||
NM_001320859.1 | Intron | NP_001307788.1 | ||||
NM_001320860.1 | Intron | NP_001307789.1 | ||||
NM_001320863.1 | Intron | NP_001307792.1 | ||||
XM_017003878.1 | Intron | XP_016859367.1 |
Set Membership: |
HapMap |