Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGATCTTATTTTCAGTCAAAGT[A/G]GTGAAGTGGCAGAAAGAAAATATCC
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 604915 | |||||||||||||||||||||||||||||
Literature Links: |
MAP3K13 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | ||||||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR - Not Available | ||||||||
AMR - Not Available |
MAP3K13 - mitogen-activated protein kinase kinase kinase 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242314.1 | Intron | NP_001229243.1 | ||||
NM_001242317.1 | Intron | NP_001229246.1 | ||||
NM_004721.4 | Intron | NP_004712.1 | ||||
XM_011513310.2 | Intron | XP_011511612.1 | ||||
XM_017007456.1 | Intron | XP_016862945.1 | ||||
XM_017007457.1 | Intron | XP_016862946.1 | ||||
XM_017007458.1 | Intron | XP_016862947.1 | ||||
XM_017007459.1 | Intron | XP_016862948.1 |
Set Membership: |
Validated |