Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTGTGGCTGCCTGCCAGTGTGCT[C/T]GCCCATGCCATCCAGGGCAGTGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607366 MIM: 609309 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HCG2040054 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HCG2040054 - uncharacterized LOC644093 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCNK12 - potassium two pore domain channel subfamily K member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022055.1 | Intron | NP_071338.1 |
MSH2 - mutS homolog 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000251.2 | Intron | NP_000242.1 | ||||
NM_001258281.1 | Intron | NP_001245210.1 | ||||
XM_005264332.3 | Intron | XP_005264389.2 | ||||
XM_011532867.1 | Intron | XP_011531169.1 |