Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTAATACCTGCATAACCTGCTCA[C/T]TCACATCTTCATTAATCAGCCTACA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 614310 | |||||||||||||||||||||||
Literature Links: |
CEP70 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CEP70 - centrosomal protein 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288964.1 | 1975 | Missense Mutation | AAT,AGT | N,S 519 | NP_001275893.1 | |
NM_001288965.1 | 1975 | Missense Mutation | AAT,AGT | N,S 517 | NP_001275894.1 | |
NM_001288966.1 | 1975 | Missense Mutation | AAT,AGT | N,S 537 | NP_001275895.1 | |
NM_001288967.1 | 1975 | Missense Mutation | AAT,AGT | N,S 385 | NP_001275896.1 | |
NM_001320598.1 | 1975 | Missense Mutation | AAT,AGT | N,S 537 | NP_001307527.1 | |
NM_001320599.1 | 1975 | Missense Mutation | AAT,AGT | N,S 537 | NP_001307528.1 | |
NM_001320600.1 | 1975 | Intron | NP_001307529.1 | |||
NM_024491.3 | 1975 | Missense Mutation | AAT,AGT | N,S 537 | NP_077817.2 | |
XM_005247805.3 | 1975 | Missense Mutation | AAT,AGT | N,S 537 | XP_005247862.1 | |
XM_011513209.2 | 1975 | Intron | XP_011511511.1 | |||
XM_017007270.1 | 1975 | Missense Mutation | AAT,AGT | N,S 537 | XP_016862759.1 | |
XM_017007271.1 | 1975 | Missense Mutation | AAT,AGT | N,S 537 | XP_016862760.1 | |
XM_017007272.1 | 1975 | Missense Mutation | AAT,AGT | N,S 537 | XP_016862761.1 | |
XM_017007273.1 | 1975 | Missense Mutation | AAT,AGT | N,S 517 | XP_016862762.1 | |
XM_017007274.1 | 1975 | Missense Mutation | AAT,AGT | N,S 517 | XP_016862763.1 | |
XM_017007275.1 | 1975 | Missense Mutation | AAT,AGT | N,S 537 | XP_016862764.1 | |
XM_017007276.1 | 1975 | Missense Mutation | AAT,AGT | N,S 419 | XP_016862765.1 | |
XM_017007277.1 | 1975 | Missense Mutation | AAT,AGT | N,S 385 | XP_016862766.1 | |
XM_017007278.1 | 1975 | Missense Mutation | AAT,AGT | N,S 361 | XP_016862767.1 | |
XM_017007279.1 | 1975 | Missense Mutation | AAT,AGT | N,S 311 | XP_016862768.1 | |
XM_017007280.1 | 1975 | Missense Mutation | AAT,AGT | N,S 278 | XP_016862769.1 | |
XM_017007281.1 | 1975 | Missense Mutation | AAT,AGT | N,S 278 | XP_016862770.1 | |
XM_017007282.1 | 1975 | Missense Mutation | AAT,AGT | N,S 278 | XP_016862771.1 | |
XM_017007283.1 | 1975 | Missense Mutation | AAT,AGT | N,S 278 | XP_016862772.1 | |
XM_017007284.1 | 1975 | Missense Mutation | AAT,AGT | N,S 278 | XP_016862773.1 |